2-Amino-5-chlorobenzyl alcohol, 98%
2-Amino-5-chlorobenzyl alcohol, 98%
2-Amino-5-chlorobenzyl alcohol, 98%
Thermo Scientific Chemicals

2-Amino-5-chlorobenzyl alcohol, 98%

CAS: 37585-25-4 | C7H8ClNO | 157.597 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H30531.06
also known as H30531-06
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
40.10
Save 5.45 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
40.10
Save 5.45 (14%)
Each
Chemical Identifiers
CAS37585-25-4
IUPAC Name(2-amino-5-chlorophenyl)methanol
Molecular FormulaC7H8ClNO
InChI KeyCLKBZWDZDVOIGJ-UHFFFAOYSA-N
SMILESNC1=C(CO)C=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)105.5-111.5?C
Appearance (Color)White to cream or pale orange
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only