6-Bromopyridine-2-carboxylic acid, 97%
6-Bromopyridine-2-carboxylic acid, 97%
6-Bromopyridine-2-carboxylic acid, 97%
Thermo Scientific Chemicals

6-Bromopyridine-2-carboxylic acid, 97%

CAS: 21190-87-4 | C6H4BrNO2 | 202.007 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H30553.03
also known as H30553-03
Price (USD)
20.65
Special offer
Online exclusive
Ends: 30-Jun-2026
24.30
Save 3.65 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
20.65
Special offer
Online exclusive
Ends: 30-Jun-2026
24.30
Save 3.65 (15%)
Each
Chemical Identifiers
CAS21190-87-4
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Sulfated Ash< 1.50%
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)> 96.0%
Assay (Silylated GC)> 96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only