Thieno[3,2-b]thiophene, 97%
Thieno[3,2-b]thiophene, 97%
Thieno[3,2-b]thiophene, 97%
Thermo Scientific Chemicals

Thieno[3,2-b]thiophene, 97%

CAS: 251-41-2 | C6H4S2 | 140.218 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H30637.03
also known as H30637-03
Price (USD)
135.65
Special offer
Online exclusive
Ends: 30-Jun-2026
159.00
Save 23.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
135.65
Special offer
Online exclusive
Ends: 30-Jun-2026
159.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS251-41-2
IUPAC Namethieno[3,2-b]thiophene
Molecular FormulaC6H4S2
InChI KeyVJYJJHQEVLEOFL-UHFFFAOYSA-N
SMILESS1C=CC2=C1C=CS2
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Melting Point (clear melt)51.0-60.0?C
Appearance (Color)White to cream to brown to gray
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only