2-Fluoro-4-methoxyphenylacetonitrile, 97%
2-Fluoro-4-methoxyphenylacetonitrile, 97%
2-Fluoro-4-methoxyphenylacetonitrile, 97%
Thermo Scientific Chemicals

2-Fluoro-4-methoxyphenylacetonitrile, 97%

CAS: 749934-29-0 | C9H8FNO | 165.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H30752.MD
also known as H30752-MD
Price (USD)
69.65
Special offer
Online exclusive
Ends: 30-Jun-2026
81.30
Save 11.65 (14%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
69.65
Special offer
Online exclusive
Ends: 30-Jun-2026
81.30
Save 11.65 (14%)
Each
Chemical Identifiers
CAS749934-29-0
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5105-1.5155 @ 20?C
FormLiquid
Appearance (Color)Clear pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only