2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H30941.06
also known as H30941-06
5 g
Catalog number H30941.06
also known as H30941-06
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS269409-99-6
IUPAC Nameethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular FormulaC15H21BO4
InChI KeyRFBZWPFBCXBBJS-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC=C1B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to yellow
Assay (GC)≥96.0%
Melting Point (clear melt)56.0-66.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only