Bromomalonaldehyde, 97%
Bromomalonaldehyde, 97%
Bromomalonaldehyde, 97%
Thermo Scientific Chemicals

Bromomalonaldehyde, 97%

CAS: 2065-75-0 | C3H3BrO2 | 150.959 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H31030.06
also known as H31030-06
Price (USD)
112.65
Special offer
Online exclusive
Ends: 30-Jun-2026
132.00
Save 19.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
112.65
Special offer
Online exclusive
Ends: 30-Jun-2026
132.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS2065-75-0
IUPAC Name2-bromopropanedial
Molecular FormulaC3H3BrO2
InChI KeySURMYNZXHKLDFO-UHFFFAOYSA-N
SMILESBrC(C=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0%
Appearance (Color)Pale yellow to brown
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only