1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene, 97%
1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene, 97%
1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene, 97%
Thermo Scientific Chemicals

1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene, 97%

CAS: 95166-77-1 | C22H20 | 284.402 g/mol
Have Questions?
Catalog NumberQuantity
H31077.03
also known as H31077-03
1 g
Catalog number H31077.03
also known as H31077-03
Price (USD)
195.65
Special offer
Online exclusive
Ends: 30-Jun-2026
230.00
Save 34.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
195.65
Special offer
Online exclusive
Ends: 30-Jun-2026
230.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS95166-77-1
IUPAC Name1-(2-phenylethenyl)-4-(2-phenylethyl)benzene
Molecular FormulaC22H20
InChI KeyVKIHNQGTZXZCPE-UHFFFAOYSA-N
SMILESC(CC1=CC=C(C=CC2=CC=CC=C2)C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)152.0-161.0?C
FormPowder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only