(2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480
(2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480
(2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480
Thermo Scientific Chemicals

(2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480

CAS: 128446-35-5 | C44H75O36 | 1180.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H31133.22
also known as H31133-22
Price (USD)
454.65
Special offer
Online exclusive
Ends: 30-Jun-2026
535.00
Save 80.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
454.65
Special offer
Online exclusive
Ends: 30-Jun-2026
535.00
Save 80.35 (15%)
Each
Chemical Identifiers
CAS128446-35-5
Molecular FormulaC44H75O36
SMILESOCC-*.[H][C@@]12O[C@H](CO)[C@@H](O[C@@]3([H])O[C@H](CO)[C@@H](O[C@@]4([H])O[C@H](CO)[C@@H](O[C@@]5([H])O[C@H](CO)[C@@H](O[C@@]6([H])O[C@H](CO)[C@@H](O[C@@]7([H])O[C@H](CO)[C@@H](O[C@@]8([H])O[C@H](CO)[C@@H](O1)[C@H](O)[C@H]8O)[C@H](O)[C@H]7O)[C@H](O)[C@H]6O)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O
Molecular Weight (g/mol)1180.05
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤7.5%
FormPowder
CommentMolar degree of Substitution (MS) 0.6-0.9
Impurity contentResidual beta-cyclodextrin ≤2.0% dry wt. basis
Appearance (Color)White
View more
2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Serio, N.; Chanthalyma, C.; Peters, S.; Levine, D.; Levine, M. 2-Hydroxypropyl beta-cyclodextrin for the enhanced performance of dual function extraction and detection systems in complex oil environments. J. Inclusion Phenom. Macrocyclic Chem. 2015, 81 (3-4), 341-346.
  2. Hernandez, D. S. G.; Silva, M. T. R.; Pardave, M. P.; Romo, M. A. R.; Avendano, C. S. Electrochemical Characterization of a 2-Hydroxypropyl b-Cyclodextrin Membrane with Tenoxicam. ECS Trans. 2015, 64 (35), 23-26.