4,4'-Di-n-nonyl-2,2'-bipyridine, 97%
4,4'-Di-n-nonyl-2,2'-bipyridine, 97%
4,4'-Di-n-nonyl-2,2'-bipyridine, 97%
Thermo Scientific Chemicals

4,4'-Di-n-nonyl-2,2'-bipyridine, 97%

CAS: 142646-58-0 | C28H44N2 | 408.674 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31476.03
also known as H31476-03
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.50
Save 9.85 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.50
Save 9.85 (15%)
Each
Chemical Identifiers
CAS142646-58-0
IUPAC Name4,4'-dinonyl-2,2'-bipyridine
Molecular FormulaC28H44N2
InChI KeyVHJFWJXYEWHCGD-UHFFFAOYSA-N
SMILESCCCCCCCCCC1=CC(=NC=C1)C1=NC=CC(CCCCCCCCC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)57.0-67.0?C
Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. M.-H. Li,; P. Keller,; J. Yang, P.-A. Albouy.An Artificial Muscle with Lamellar Structure Based on a Nematic Triblock Copolymer. Journal name. 2004, 16e (21),1922-1925.
  2. Jian Qiu,; Krzysztof Matyjaszewski,; Laurent Thouin,; Christian Amatore. Cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization . Macromolecular Chemistry and Physics. 2000, 201 (14),1625-1631.