2-Aminobenzothiazole-6-carboxylic acid, 95%
2-Aminobenzothiazole-6-carboxylic acid, 95%
2-Aminobenzothiazole-6-carboxylic acid, 95%
2-Aminobenzothiazole-6-carboxylic acid, 95%
Thermo Scientific Chemicals

2-Aminobenzothiazole-6-carboxylic acid, 95%

CAS: 93-85-6 | C8H6N2O2S | 194.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31512.06
also known as H31512-06
Price (USD)
237.65
Online Exclusive
264.00
Save 26.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
237.65
Online Exclusive
264.00
Save 26.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS93-85-6
IUPAC Name2-amino-1,3-benzothiazole-6-carboxylic acid
Molecular FormulaC8H6N2O2S
InChI KeyZEAKWWWXCZMODH-UHFFFAOYSA-N
SMILESNC1=NC2=CC=C(C=C2S1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Yellow
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only