Tri-n-butyl(1-ethoxyvinyl)tin, 95%
Tri-n-butyl(1-ethoxyvinyl)tin, 95%
Tri-n-butyl(1-ethoxyvinyl)tin, 95%
Thermo Scientific Chemicals

Tri-n-butyl(1-ethoxyvinyl)tin, 95%

CAS: 97674-02-7 | C16H34OSn | 361.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H31562.03
also known as H31562-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS97674-02-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4720-1.4800 @ 20?C
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only