4-(4-Bromophenyl)imidazole, 97%
4-(4-Bromophenyl)imidazole, 97%
4-(4-Bromophenyl)imidazole, 97%
Thermo Scientific Chemicals

4-(4-Bromophenyl)imidazole, 97%

CAS: 13569-96-5 | C9H7BrN2 | 223.073 g/mol
Have Questions?
Catalog NumberQuantity
H31581.06
also known as H31581-06
5 g
Catalog number H31581.06
also known as H31581-06
Price (USD)
79.65
Online Exclusive
88.50
Save 8.85 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
79.65
Online Exclusive
88.50
Save 8.85 (10%)
Each
Chemical Identifiers
CAS13569-96-5
IUPAC Name5-(4-bromophenyl)-1H-imidazole
Molecular FormulaC9H7BrN2
InChI KeyYLIOAWKNPLJMID-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)C1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)137.0-143.0?C
Appearance (Color)Cream
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only