4-Fluorophthalic acid, 98%
4-Fluorophthalic acid, 98%
4-Fluorophthalic acid, 98%
Thermo Scientific Chemicals

4-Fluorophthalic acid, 98%

CAS: 320-97-8 | C8H5FO4 | 184.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H31597.09
also known as H31597-09
Price (USD)
226.65
Online Exclusive
252.00
Save 25.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
226.65
Online Exclusive
252.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS320-97-8
IUPAC Name4-fluorobenzene-1,2-dicarboxylic acid
Molecular FormulaC8H5FO4
InChI KeyOMCXTFVBNCFZMY-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(F)C=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
4-Fluorophthalic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophthalic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. M Grifoll; S A Selifonov and P J Chapman. Transformation of Substituted Fluorenes and Fluorene Analogs by Pseudomonas sp. Strain F274. Appl. Environ. Microbiol. 1995, 61 (9), 3490-3493.
  2. F. J. Williams; P. E. Donahue. Reaction of thiophenoxides with nitro- and halo-substituted phthalic anhydrides. J. Org. Chem. 1989, 43 (2), 255-258.