2-Bromo-6-methoxybenzoic acid, 98%
2-Bromo-6-methoxybenzoic acid, 98%
2-Bromo-6-methoxybenzoic acid, 98%
2-Bromo-6-methoxybenzoic acid, 98%
Thermo Scientific Chemicals

2-Bromo-6-methoxybenzoic acid, 98%

CAS: 31786-45-5 | C8H7BrO3 | 231.045 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31604.03
also known as H31604-03
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.60
Save 12.95 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.60
Save 12.95 (15%)
Each
Add to cart
Chemical Identifiers
CAS31786-45-5
IUPAC Name2-bromo-6-methoxybenzoic acid
Molecular FormulaC8H7BrO3
InChI KeyBNYFDICJYCBPRW-UHFFFAOYSA-N
SMILESCOC1=C(C(O)=O)C(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)123.0-128.0?C
Appearance (Color)Pale cream to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
FormPowder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only