3,5-Difluoro-4-hydroxybenzonitrile, 97%
3,5-Difluoro-4-hydroxybenzonitrile, 97%
3,5-Difluoro-4-hydroxybenzonitrile, 97%
3,5-Difluoro-4-hydroxybenzonitrile, 97%
Thermo Scientific Chemicals

3,5-Difluoro-4-hydroxybenzonitrile, 97%

CAS: 2967-54-6 | C7H3F2NO | 155.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H31620.MD
also known as H31620-MD
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.90
Save 13.25 (15%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.90
Save 13.25 (15%)
Each
Add to cart
Chemical Identifiers
CAS2967-54-6
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)198.0-208.0?C
Assay (GC)≥96.0%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only