4-(4-Bromophenoxy)benzaldehyde, 97%
4-(4-Bromophenoxy)benzaldehyde, 97%
4-(4-Bromophenoxy)benzaldehyde, 97%
4-(4-Bromophenoxy)benzaldehyde, 97%
Thermo Scientific Chemicals

4-(4-Bromophenoxy)benzaldehyde, 97%

CAS: 69240-56-8 | C13H9BrO2 | 277.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31702.06
also known as H31702-06
Price (USD)
251.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
251.00
Each
Add to cart
Chemical Identifiers
CAS69240-56-8
IUPAC Name4-(4-bromophenoxy)benzaldehyde
Molecular FormulaC13H9BrO2
InChI KeyWMDDJOBNAOVSLP-UHFFFAOYSA-N
SMILESBrC1=CC=C(OC2=CC=C(C=O)C=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Free acid (titration)≤1.5%
Melting Point (clear melt)68.0-74.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only