2,4-Dimethoxybenzenesulfonamide, 96%
2,4-Dimethoxybenzenesulfonamide, 96%
2,4-Dimethoxybenzenesulfonamide, 96%
2,4-Dimethoxybenzenesulfonamide, 96%
Thermo Scientific Chemicals

2,4-Dimethoxybenzenesulfonamide, 96%

CAS: 51770-71-9 | C8H11NO4S | 217.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31707.03
also known as H31707-03
Price (USD)
48.65
Online Exclusive
53.40
Save 4.75 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
48.65
Online Exclusive
53.40
Save 4.75 (9%)
Each
Add to cart
Chemical Identifiers
CAS51770-71-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)163.5-172.5?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only