5-Hydroxy-2-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
5-Hydroxy-2-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
5-Hydroxy-2-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Hydroxy-2-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H31734.03
also known as H31734-03
1 g
Catalog number H31734.03
also known as H31734-03
Price (USD)
78.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.90
Save 13.25 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
78.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.90
Save 13.25 (14%)
Each
Chemical Identifiers
CAS216766-12-0
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Yellow to pale brown
Assay (GC)≥96.0%
Melting Point (clear melt)169.0-179.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only