9-Formyl-8-hydroxyjulolidine, 97%
9-Formyl-8-hydroxyjulolidine, 97%
9-Formyl-8-hydroxyjulolidine, 97%
Thermo Scientific Chemicals

9-Formyl-8-hydroxyjulolidine, 97%

CAS: 63149-33-7 | C13H15NO2 | 217.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31782.06
also known as H31782-06
Price (USD)
223.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
223.00
Each
Chemical Identifiers
CAS63149-33-7
IUPAC Name8-hydroxy-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene-7-carbaldehyde
Molecular FormulaC13H15NO2
InChI KeyNRZXBDYODHLZBF-UHFFFAOYSA-N
SMILESOC1=C(C=O)C=C2CCCN3CCCC1=C23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale green
Assay (GC)≥96.0%
Melting Point (clear melt)67.0-77.0?C
FormPowder
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only