Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines
Solubility
Reacts with water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
RUO – Research Use Only
General References:
- Min Shi.; Jian-Kang Jiang.; Yan-Shu Feng. Titanium(IV) Chloride and the Amine-Promoted Baylis-Hillman Reaction. Org. Lett. 2000, 16, (2), 2397-2400.
- Min Shi.; Yan-Shu Feng. Titanium(IV) Chloride and Quaternary Ammonium Salt Promoted Baylis-Hillman Reaction. J. Org. Chem. 2001, 66, (2), 406-411.