4-Bromo-2-methylbenzenesulfonyl chloride, 97%
4-Bromo-2-methylbenzenesulfonyl chloride, 97%
4-Bromo-2-methylbenzenesulfonyl chloride, 97%
4-Bromo-2-methylbenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

4-Bromo-2-methylbenzenesulfonyl chloride, 97%

CAS: 139937-37-4 | C7H6BrClO2S | 269.537 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H31835.09
also known as H31835-09
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS139937-37-4
IUPAC Name4-bromo-2-methylbenzene-1-sulfonyl chloride
Molecular FormulaC7H6BrClO2S
InChI KeyKNBSFUSBZNMAKU-UHFFFAOYSA-N
SMILESCC1=C(C=CC(Br)=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)59.0-68.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only