5-Chloro-2-methoxybenzaldehyde, 97%
5-Chloro-2-methoxybenzaldehyde, 97%
5-Chloro-2-methoxybenzaldehyde, 97%
5-Chloro-2-methoxybenzaldehyde, 97%
Thermo Scientific Chemicals

5-Chloro-2-methoxybenzaldehyde, 97%

CAS: 7035-09-8 | C8H7ClO2 | 170.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H31860.14
also known as H31860-14
Price (USD)
252.65
Special offer
Online exclusive
Ends: 30-Jun-2026
297.00
Save 44.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
252.65
Special offer
Online exclusive
Ends: 30-Jun-2026
297.00
Save 44.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7035-09-8
IUPAC Name5-chloro-2-methoxybenzaldehyde
Molecular FormulaC8H7ClO2
InChI KeyHXTWDGGMXZXOIV-UHFFFAOYSA-N
SMILESCOC1=C(C=O)C=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)73.0-83.0?C
Free acid (titration)≤1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only