6-Aminopyridine-2-carboxylic acid, 95%
6-Aminopyridine-2-carboxylic acid, 95%
6-Aminopyridine-2-carboxylic acid, 95%
6-Aminopyridine-2-carboxylic acid, 95%
Thermo Scientific Chemicals

6-Aminopyridine-2-carboxylic acid, 95%

CAS: 23628-31-1 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H31861.09
also known as H31861-09
Price (USD)
279.65
Online Exclusive
310.00
Save 30.35 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
279.65
Online Exclusive
310.00
Save 30.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS23628-31-1
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥94.0%
FormPowder
Appearance (Color)Pale cream
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

Solubility
Soluble in hot water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only
Abdullah Zafar.; Steven J. Geib.; Yoshitomo Hamuro.; Andrew J. Carr.; Andrew D. Hamilton. Hydrogen Bonding Control of Molecular Self-Assembly: Aggregation Behavior of Acylaminopyridine-Carboxylic Acid Derivatives in Solution and the Solid State.Tetrahedron. 2000, 56 (43),8419-8427 .