Benzothiazole-6-carboxylic acid, 96%
Benzothiazole-6-carboxylic acid, 96%
Benzothiazole-6-carboxylic acid, 96%
Thermo Scientific Chemicals

Benzothiazole-6-carboxylic acid, 96%

CAS: 3622-35-3 | C8H5NO2S | 179.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H31868.03
also known as H31868-03
Price (USD)
49.65
Online exclusive
55.50
Save 5.85
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
49.65
Online exclusive
55.50
Save 5.85
Each
Chemical Identifiers
CAS3622-35-3
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White to cream
FormPowder
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (HPLC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only