Sulfuryl chloride, 1M soln. in dichloromethane
Sulfuryl chloride, 1M soln. in dichloromethane
Sulfuryl chloride, 1M soln. in dichloromethane
Thermo Scientific Chemicals

Sulfuryl chloride, 1M soln. in dichloromethane

CAS: 7791-25-5 | Cl2O2S | 134.96 g/mol
Have Questions?
Catalog NumberQuantity
H31899.AP
also known as H31899-AP
500 mL
Catalog number H31899.AP
also known as H31899-AP
Price (USD)
181.65
Special offer
Online exclusive
Ends: 30-Jun-2026
214.00
Save 32.35 (15%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
181.65
Special offer
Online exclusive
Ends: 30-Jun-2026
214.00
Save 32.35 (15%)
Each
Chemical Identifiers
CAS7791-25-5
IUPAC Namesulfuroyl dichloride
Molecular FormulaCl2O2S
InChI KeyYBBRCQOCSYXUOC-UHFFFAOYSA-N
SMILESClS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (Titration ex Chloride)0.90-1.20M
Sulfuryl chloride is often used as a source of chlorine. is widely used as a reagent in the conversion of C-H ? C-Cl adjacent to activating substituent such as carbonyls and sulfoxides. It also chlorinates alkanes, alkenes, alkynes, aromatics, ethers (such as tetrahydrofuran) and epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfuryl chloride is often used as a source of chlorine. is widely used as a reagent in the conversion of C-H → C-Cl adjacent to activating substituent such as carbonyls and sulfoxides. It also chlorinates alkanes, alkenes, alkynes,
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Joo-Hack Youn.; Rudolf Herrmann. A simple and efficient preparation of sulfinyl chlorides from disulfides and sulfuryl chloride.Tetrahedron Lett. 1986, 27 (13), 1493-1494 .