6-Fluorochromone, 97%
6-Fluorochromone, 97%
6-Fluorochromone, 97%
Thermo Scientific Chemicals

6-Fluorochromone, 97%

CAS: 105300-38-7 | C9H5FO2 | 164.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31907.03
also known as H31907-03
Price (USD)
33.65
Online Exclusive
37.40
Save 3.75 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
33.65
Online Exclusive
37.40
Save 3.75 (10%)
Each
Chemical Identifiers
CAS105300-38-7
IUPAC Name6-fluoro-4H-chromen-4-one
Molecular FormulaC9H5FO2
InChI KeyWHIGSYZUTMYUAX-UHFFFAOYSA-N
SMILESFC1=CC=C2OC=CC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)163.0-172.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only