6-Bromothiochroman-4-one, 95%
6-Bromothiochroman-4-one, 95%
6-Bromothiochroman-4-one, 95%
6-Bromothiochroman-4-one, 95%
Thermo Scientific Chemicals

6-Bromothiochroman-4-one, 95%

CAS: 13735-13-2 | C9H7BrOS | 243.118 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31968.03
also known as H31968-03
Price (USD)
72.65
Online Exclusive
80.30
Save 7.65 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.30
Save 7.65 (10%)
Each
Add to cart
Chemical Identifiers
CAS13735-13-2
IUPAC Name6-bromo-3,4-dihydro-2H-1-benzothiopyran-4-one
Molecular FormulaC9H7BrOS
InChI KeySBNPYDJTVGRDAA-UHFFFAOYSA-N
SMILESBrC1=CC2=C(SCCC2=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream
Melting Point (clear melt)66.5-75.5?C
Assay (GC)≥94.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only
Brett M. Johnson.; Malcolm P. Huestis. Sequential C3 and C5 Direct C-H Arylation of Imidazo[1,2-a]pyrazines with (Hetero)aryl Bromides.Eur. J. Org. Chem. 2014, (8),1589-1593 .