4-n-Dodecylbenzoic acid, 98%
4-n-Dodecylbenzoic acid, 98%
4-n-Dodecylbenzoic acid, 98%
Thermo Scientific Chemicals

4-n-Dodecylbenzoic acid, 98%

CAS: 21021-55-6 | C19H30O2 | 290.447 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31983.06
also known as H31983-06
Price (USD)
189.65
Online Exclusive
210.00
Save 20.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
189.65
Online Exclusive
210.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS21021-55-6
IUPAC Name4-dodecylbenzoic acid
Molecular FormulaC19H30O2
InChI KeyJCHCIOJFCCQEKZ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only