2-Furaldehyde 2,2-dimethylhydrazone, 98%
2-Furaldehyde 2,2-dimethylhydrazone, 98%
2-Furaldehyde 2,2-dimethylhydrazone, 98%
Thermo Scientific Chemicals

2-Furaldehyde 2,2-dimethylhydrazone, 98%

CAS: 14064-21-2 | C7H10N2O | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number H32013.18
also known as H32013-18
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS14064-21-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to dark red-brown
Assay (GC)≥97.5%
Refractive Index1.5765-1.5805 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only