2-Chloro-4-methoxybenzoic acid, 98%
2-Chloro-4-methoxybenzoic acid, 98%
2-Chloro-4-methoxybenzoic acid, 98%
Thermo Scientific Chemicals

2-Chloro-4-methoxybenzoic acid, 98%

CAS: 21971-21-1 | C8H7ClO3 | 186.591 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32014.06
also known as H32014-06
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS21971-21-1
IUPAC Name2-chloro-4-methoxybenzoic acid
Molecular FormulaC8H7ClO3
InChI KeyIBANGHTVBPZCHF-UHFFFAOYSA-N
SMILESCOC1=CC(Cl)=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)208.0-214.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only