Potassium phenoxymethyltrifluoroborate, 95%, Thermo Scientific Chemicals
Potassium phenoxymethyltrifluoroborate, 95%, Thermo Scientific Chemicals
Potassium phenoxymethyltrifluoroborate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium phenoxymethyltrifluoroborate, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H32030.03
also known as H32030-03
1 g
Catalog number H32030.03
also known as H32030-03
Price (USD)
238.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
238.00
Each
Chemical Identifiers
CAS1027642-30-3
IUPAC Namepotassium trifluoro(phenoxymethyl)boranuide
Molecular FormulaC7H7BF3KO
InChI KeyOVYSNXCRBZPJIG-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)COC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Proton NMRConforms to structure
Assay (HPLC)≥95.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only