2,2,2-Trimethylthioacetamide, 97%
2,2,2-Trimethylthioacetamide, 97%
2,2,2-Trimethylthioacetamide, 97%
Thermo Scientific Chemicals

2,2,2-Trimethylthioacetamide, 97%

CAS: 630-22-8 | C5H11NS | 117.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H32092.03
also known as H32092-03
Price (USD)
55.65
Online Exclusive
61.70
Save 6.05 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.70
Save 6.05 (10%)
Each
Chemical Identifiers
CAS630-22-8
IUPAC Name2,2-dimethylpropanethioamide
Molecular FormulaC5H11NS
InChI KeyFJZJUSOFGBXHCV-UHFFFAOYSA-N
SMILESCC(C)(C)C(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)112.0-121.0?C
2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

Solubility
Soluble in water (10 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. J Taltavull.; J Serrat, J Gracia.; A Gavalda. Synthesis and Biological Activity of Pyrido [3', 2': 4, 5] thieno [3, 2-d] pyrimidines as Phosphodiesterase Type 4 Inhibitors. J. Med. Chem. 201053 (19), 6912-6922.
  2. JF Cavalla.; NE Webb.; JAD Willis. 3-Amino-4-thiocarbamoyl-3-pyrrolines: their conversion into pyrrolo [3, 4-d] pyrimidine-4-thiols. J. Chem. Soc. C. 19671 (0), 698-701.