2-Fluoro-4-methoxybenzyl alcohol, 97%
2-Fluoro-4-methoxybenzyl alcohol, 97%
2-Fluoro-4-methoxybenzyl alcohol, 97%
2-Fluoro-4-methoxybenzyl alcohol, 97%
Thermo Scientific Chemicals

2-Fluoro-4-methoxybenzyl alcohol, 97%

CAS: 405-09-4 | C8H9FO2 | 156.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H32118.03
also known as H32118-03
Price (USD)
68.65
Online Exclusive
76.30
Save 7.65 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
68.65
Online Exclusive
76.30
Save 7.65 (10%)
Each
Add to cart
Chemical Identifiers
CAS405-09-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.5225-1.5275 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only