11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide, 95%
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide, 95%
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide, 95%
Thermo Scientific Chemicals

11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide, 95%

CAS: 207399-07-3 | C36H44ClIN2 | 667.116 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32150.03
also known as H32150-03
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS207399-07-3
IUPAC Name2-(2-{2-chloro-3-[2-(3,3-dimethyl-1-propyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-3,3-dimethyl-1-propyl-3H-indol-1-ium iodide
Molecular FormulaC36H44ClIN2
InChI KeyIRPKBYJYVJOQHQ-UHFFFAOYSA-M
SMILES[I-].CCCN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](CCC)C3=CC=CC=C3C2(C)C)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis(Iodine) : 18.52-19.52%
Appearance (Color)Green/gold to dark green
Assay (unspecified)(by Iodine EA) : >94.0 to<106.0%
lambda-max (UV-Vis)(in Ethanol) : 782-786 nm
FormCrystals or crystalline powder
View more
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide is used as a laser and IR dye. It is also used as lithographic computer to plate laser applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Chloro-1,1′-di-n-propyl-3,3,3′,3′-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide is used as a laser and IR dye. It is also used as lithographic computer to plate laser applications.

Solubility
It is soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only