2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, 99%
2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, 99%
2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, 99%
Thermo Scientific Chemicals

2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, 99%

CAS: 4359-46-0 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32187.06
also known as H32187-06
Price (USD)
114.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
114.00
Each
Chemical Identifiers
CAS4359-46-0
IUPAC Name2-ethyl-4-methyl-1,3-dioxolane
Molecular FormulaC6H12O2
InChI KeyCSZCLQLJVFLXLI-UHFFFAOYNA-N
SMILESCCC1OCC(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless
Refractive Index1.4025-1.4055 @ 20?C
FormLiquid
2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, is used as a flavouring agent in food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, is used as a flavouring agent in food industry.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. RJ Kern. Twelve-membered polyether rings. Cyclic tetramers of some olefin oxides. The Journal of Organic Chemistry. 196833 (1), 388-390.
  2. BE Leggetter.; RK Brown. Hydrogenolysis by alcl3-lialh4 of ether solutions of cis and trans isomers of 2, 4-disubstituted-1, 3-dioxolanes. Canadian Journal of Chemistry. 196543 (5), 1030-1035.