2,6-Difluoro-3-hydroxybenzaldehyde, 95%
2,6-Difluoro-3-hydroxybenzaldehyde, 95%
2,6-Difluoro-3-hydroxybenzaldehyde, 95%
Thermo Scientific Chemicals

2,6-Difluoro-3-hydroxybenzaldehyde, 95%

CAS: 152434-88-3 | C7H4F2O2 | 158.104 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H32250.MD
also known as H32250-MD
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS152434-88-3
IUPAC Name2,6-difluoro-3-hydroxybenzaldehyde
Molecular FormulaC7H4F2O2
InChI KeyNHGSYGIEJAONJB-UHFFFAOYSA-N
SMILESOC1=C(F)C(C=O)=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to pale brown
FormPowder
Assay (GC)≥94.0%
Melting Point (clear melt)106.5-118.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only