Cyclopropanethiocarboxamide, 97%
Cyclopropanethiocarboxamide, 97%
Cyclopropanethiocarboxamide, 97%
Thermo Scientific Chemicals

Cyclopropanethiocarboxamide, 97%

CAS: 20295-34-5 | C4H7NS | 101.167 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32315.03
also known as H32315-03
Price (USD)
86.65
Special offer
Online exclusive
Ends: 28-Dec-2026
102.00
Save 15.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
Ends: 28-Dec-2026
102.00
Save 15.35
Each
Chemical Identifiers
CAS20295-34-5
IUPAC Namecyclopropanecarbothioamide
Molecular FormulaC4H7NS
InChI KeyIIPJWNFOLPDTEQ-UHFFFAOYSA-N
SMILESNC(=S)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)110.0-119.0°C
Cyclopropanecarbothioamide is used for purification in single-step preparation of thiazolo [5, 4-b] pyridine-and thiazolo [5, 4-c] pyridine derivatives from chloronitropyridines and thioamides, or thioureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarbothioamide is used for purification in single-step preparation of thiazolo [5, 4-b] pyridine-and thiazolo [5, 4-c] pyridine derivatives from chloronitropyridines and thioamides, or thioureas.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kiran P. Sahasrabudhe,; M. Angels Estiarte,; Darlene Tan;, Sheila Zipfel,; Matthew Cox,; Donogh J. R. O'Mahony,; William T. Edwards,; Matthew A. J. Duncton†. A single-step preparation of thiazolo[5,4-b]pyridine- and thiazolo[5,4-c]pyridine derivatives from chloronitropyridines and thioamides, or thioureas. Journal of Heterocyclic Chemistry. 2009, 46(6),1125-1131.
  2. authors. title . Journal name. year of publication , volume issue , pg no.