2-Chloro-N-methylacetamide, 97%
2-Chloro-N-methylacetamide, 97%
2-Chloro-N-methylacetamide, 97%
Thermo Scientific Chemicals

2-Chloro-N-methylacetamide, 97%

CAS: 96-30-0 | C3H6ClNO | 107.537 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32362.06
also known as H32362-06
Price (USD)
74.65
Online Exclusive
82.70
Save 8.05 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
74.65
Online Exclusive
82.70
Save 8.05 (10%)
Each
Chemical Identifiers
CAS96-30-0
IUPAC Name2-chloro-N-methylacetamide
Molecular FormulaC3H6ClNO
InChI KeyHOZLOOPIXHWKCI-UHFFFAOYSA-N
SMILESCNC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
Melting Point (clear melt)38.0-47.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only