1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester, 95%
1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester, 95%
1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester, 95%
Thermo Scientific Chemicals

1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester, 95%

CAS: 1029684-37-4 | C14H23BN2O3 | 278.159 g/mol
Have Questions?
Catalog NumberQuantity
H32369.ME
also known as H32369-ME
500 mg
Catalog number H32369.ME
also known as H32369-ME
Price (USD)
692.65
Online Exclusive
769.00
Save 76.35 (10%)
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
692.65
Online Exclusive
769.00
Save 76.35 (10%)
Each
Chemical Identifiers
CAS1029684-37-4
IUPAC Name1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole
Molecular FormulaC14H23BN2O3
InChI KeyBVIWGBKKGPHNAB-UHFFFAOYNA-N
SMILESCC1(C)OB(OC1(C)C)C1=CN=CN1C1CCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (GC)≥94.0%
FormPowder
Melting Point (clear melt)121.0-133.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only