2,6-Dichloro-4-(4-fluorobenzoyl)pyrimidine, 95%, Thermo Scientific Chemicals
2,6-Dichloro-4-(4-fluorobenzoyl)pyrimidine, 95%, Thermo Scientific Chemicals
2,6-Dichloro-4-(4-fluorobenzoyl)pyrimidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dichloro-4-(4-fluorobenzoyl)pyrimidine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H32374.MD
also known as H32374-MD
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.20
Save 12.55 (14%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.20
Save 12.55 (14%)
Each
Chemical Identifiers
CAS1099597-81-5
IUPAC Name2,4-dichloro-6-(4-fluorobenzoyl)pyrimidine
Molecular FormulaC11H5Cl2FN2O
InChI KeyQIULZDCQGAXJIB-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C(=O)C1=NC(Cl)=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥94.0%
Appearance (Color)Pale yellow
Melting Point (clear melt)91.0-104.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only