2-Chloro-6-fluorobenzothiazole, 98%
2-Chloro-6-fluorobenzothiazole, 98%
2-Chloro-6-fluorobenzothiazole, 98%
Thermo Scientific Chemicals

2-Chloro-6-fluorobenzothiazole, 98%

CAS: 399-74-6 | C7H3ClFNS | 187.616 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32383.03
also known as H32383-03
Price (USD)
77.65
Online Exclusive
85.60
Save 7.95 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
77.65
Online Exclusive
85.60
Save 7.95 (9%)
Each
Chemical Identifiers
CAS399-74-6
IUPAC Name2-chloro-6-fluoro-1,3-benzothiazole
Molecular FormulaC7H3ClFNS
InChI KeyISIIQFDYFMPPOA-UHFFFAOYSA-N
SMILESFC1=CC=C2N=C(Cl)SC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream
Assay (GC)≥97.5%
Melting Point (clear melt)89.0-96.0?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only