As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.
Solubility
Sparingly Soluble (0.40 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
- Yi-Che Su,; Feng-Chih Chang. Synthesis and characterization of fluorinated polybenzoxazine material with low dielectric constant. Polymer. 2003, 44 (26), 7989-7996
- Fraser Hof Dr.,; Denise M. Scofield,; W. Bernd Schweizer Dr; François Diederich. A Weak Attractive Interaction between Organic Fluorine and an Amide Grou†. Angewandte Chemie . 2004, 43 (38), 5056-5059.