4-Amino-3-(trifluoromethyl)benzoic acid, 98%
4-Amino-3-(trifluoromethyl)benzoic acid, 98%
4-Amino-3-(trifluoromethyl)benzoic acid, 98%
4-Amino-3-(trifluoromethyl)benzoic acid, 98%
Thermo Scientific Chemicals

4-Amino-3-(trifluoromethyl)benzoic acid, 98%

CAS: 400-76-0 | C8H6F3NO2 | 205.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H32487.09
also known as H32487-09
Price (USD)
112.65
Special offer
Online exclusive
Ends: 30-Jun-2026
132.00
Save 19.35 (15%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
112.65
Special offer
Online exclusive
Ends: 30-Jun-2026
132.00
Save 19.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS400-76-0
IUPAC Name4-amino-3-(trifluoromethyl)benzoic acid
Molecular FormulaC8H6F3NO2
InChI KeyNPPPORJZPNJXNQ-UHFFFAOYSA-N
SMILESNC1=C(C=C(C=C1)C(O)=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)210.0-216.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only