2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile, 97%
2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile, 97%
2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile, 97%
2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile, 97%
Thermo Scientific Chemicals

2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile, 97%

CAS: 4651-91-6 | C9H10N2S | 178.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H32544.03
also known as H32544-03
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
37.80
Save 5.15 (14%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
37.80
Save 5.15 (14%)
Each
Add to cart
Chemical Identifiers
CAS4651-91-6
IUPAC Name2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
Molecular FormulaC9H10N2S
InChI KeyADHVMGAFAKSNOM-UHFFFAOYSA-N
SMILESNC1=C(C#N)C2=C(CCCC2)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)143.0-152.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only