5-Phenyl-1H-pyrazole-4-carboxylic acid, 97%
5-Phenyl-1H-pyrazole-4-carboxylic acid, 97%
5-Phenyl-1H-pyrazole-4-carboxylic acid, 97%
Thermo Scientific Chemicals

5-Phenyl-1H-pyrazole-4-carboxylic acid, 97%

CAS: 5504-65-4 | C10H8N2O2 | 188.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H32560.03
also known as H32560-03
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS5504-65-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only