Ethyl 2-bromo-4,4,4-trifluorobutyrate, 97%
Ethyl 2-bromo-4,4,4-trifluorobutyrate, 97%
Ethyl 2-bromo-4,4,4-trifluorobutyrate, 97%
Thermo Scientific Chemicals

Ethyl 2-bromo-4,4,4-trifluorobutyrate, 97%

CAS: 367-33-9 | C6H8BrF3O2 | 249.027 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H32572.MD
also known as H32572-MD
Price (USD)
59.65
Special offer
Online exclusive
Ends: 30-Jun-2026
69.90
Save 10.25 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
59.65
Special offer
Online exclusive
Ends: 30-Jun-2026
69.90
Save 10.25 (15%)
Each
Chemical Identifiers
CAS367-33-9
IUPAC Nameethyl 2-bromo-4,4,4-trifluorobutanoate
Molecular FormulaC6H8BrF3O2
InChI KeyPOVYUFTVLGOXEE-UHFFFAOYNA-N
SMILESCCOC(=O)C(Br)CC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3960-1.4010 @ 20?C
Ethyl 2-bromo-4,4,4-trifluorobutyrate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-bromo-4,4,4-trifluorobutyrate is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Pin Wang.; Yi Tang.; David A. Incorporation of Trifluoroisoleucine into Proteins in Vivo. J. Am. Chem. Soc. 2003, 125 (23), 6900-6906.
  2. H. M. Walborsky.; M. Baum.; D. F. Loncrini. The Syntheses of ι-Trifluoromethyl Amino Acids. II.1,2 Their Microbiological Activities. J. Am. Chem. Soc. 1955, 77 (13), 3637-3640.