Bis(4-fluorophenyl) disulfide, 98%
Bis(4-fluorophenyl) disulfide, 98%
Bis(4-fluorophenyl) disulfide, 98%
Thermo Scientific Chemicals

Bis(4-fluorophenyl) disulfide, 98%

CAS: 405-31-2 | C12H8F2S2 | 254.313 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32589.06
also known as H32589-06
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS405-31-2
IUPAC Name1-fluoro-4-[(4-fluorophenyl)disulfanyl]benzene
Molecular FormulaC12H8F2S2
InChI KeySLXZJIKDNHDPKL-UHFFFAOYSA-N
SMILESFC1=CC=C(SSC2=CC=C(F)C=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.6125-1.6165 @ 20?C
Bis(4-fluorophenyl) disulfide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-fluorophenyl) disulfide is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Judith Giordan.; Hans Bock. Radical ions, 51. Oxidative Rearrangement of Diphenyl Disulfides to Thianthrenes. Chemical Communications. 1982, 115 (7), 2548-2559.
  2. Z. Y. Wang.; G. Bonanno.; A. S. Hay. Novel redox reactions between diaryl disulfides and iodoarenes. J. Org. Chem. 1992, 57 (9), 2751-2753.