4-Bromo-2-(trifluoromethoxy)anisole, 98%
4-Bromo-2-(trifluoromethoxy)anisole, 98%
4-Bromo-2-(trifluoromethoxy)anisole, 98%
Thermo Scientific Chemicals

4-Bromo-2-(trifluoromethoxy)anisole, 98%

CAS: 853771-88-7 | C8H6BrF3O2 | 271.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32645.06
also known as H32645-06
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS853771-88-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4800-1.4840 @ 20?C
4-Bromo-2-(trifluoromethoxy)anisole employed in biological evaluation of some analogs of the antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-(trifluoromethoxy)anisole employed in biological evaluation of some analogs of the antitumor agents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Frédéric R. Leroux,; Baptiste Manteau,; Jean-Pierre Vors,; Sergiy Pazenok. Trifluoromethyl ethers - synthesis and properties of an unusual substituent. J. Org. Chem. 2008, 4 (13)
  2. Stuart T. Hazeldine,; Lisa Polin,; Juiwanna Kushner,; Kathryn White,; Thomas H. Corbett,; Jason Biehl,; Jerome P. Horwitz. Synthesis and biological evaluation of some analogs of the antitumor agents, 2-{4-[(7-chloro-2-quinoxalinyl)oxy]phenoxy}propionic acid, and 2-{4-[(7-bromo-2-quinolinyl)oxy]phenoxy}propionic acid. Bioorganic & Medicinal Chemistry. 2005, 13(4), 1069-1081.