4,5-Difluorosalicylaldehyde, 98%
4,5-Difluorosalicylaldehyde, 98%
4,5-Difluorosalicylaldehyde, 98%
Thermo Scientific Chemicals

4,5-Difluorosalicylaldehyde, 98%

CAS: 199287-52-0 | C7H4F2O2 | 158.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H32680.MD
also known as H32680-MD
Price (USD)
66.65
Online Exclusive
74.30
Save 7.65 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
74.30
Save 7.65 (10%)
Each
Chemical Identifiers
CAS199287-52-0
IUPAC Name4,5-difluoro-2-hydroxybenzaldehyde
Molecular FormulaC7H4F2O2
InChI KeyHRLWPXGNZQEUHM-UHFFFAOYSA-N
SMILESOC1=CC(F)=C(F)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown or pale pink
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)68.0-74.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only