1,2,4-Triazolo[4,3-a]pyridine-3-thiol, 96%
1,2,4-Triazolo[4,3-a]pyridine-3-thiol, 96%
1,2,4-Triazolo[4,3-a]pyridine-3-thiol, 96%
1,2,4-Triazolo[4,3-a]pyridine-3-thiol, 96%
Thermo Scientific Chemicals

1,2,4-Triazolo[4,3-a]pyridine-3-thiol, 96%

CAS: 6952-68-7 | C6H5N3S | 151.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32694.03
also known as H32694-03
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.90
Save 7.25 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.90
Save 7.25 (15%)
Each
Add to cart
Chemical Identifiers
CAS6952-68-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥95.0%
Appearance (Color)White to pale cream
Melting Point (clear melt)210.0-219.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only